MilliporeSigma
  • Influence of Effective Water-to-Cement Ratios on Internal Damage and Salt Scaling of Concrete with Superabsorbent Polymer.
Influence of Effective Water-to-Cement Ratios on Internal Damage and Salt Scaling of Concrete with Superabsorbent Polymer.
Materials (Basel, Switzerland) (2019-11-27)
Minsoo Kim, Sung-Hoon Kang, Sung-Gul Hong, Juhyuk Moon
ABSTRACT
Superabsorbent polymer (SAP) is attracting attention as a water-entraining admixture that reduces shrinkage or heals cracks in concrete. Cross-linked sodium polyacrylate SAPs, which are the most widely produced SAPs in the global market, are applicable as concrete admixtures. However, there have been contradictory results on the freeze-thaw resistance of concrete with SAPs. This study aims to clarify these results considering the water absorption behavior of SAPs in hardened concrete when effective water-to-cement ratios are different. Firstly, the absorbencies of one kind of cross-linked sodium polyacrylate SAP (SAP_SP) in pore solution and fresh mortar were measured by a tea bag test and flow test, respectively. Pore size distribution, capillary water absorption, and deformation during freeze-thaw cycles were analyzed for mortar samples with varying SAP_SP dosages. In the main tests, concrete samples with three different SAP_SPs/cement ratios (0.1%, 0.2%, and 0.3%) and a reference sample were prepared, and internal damage and salt scaling were measured under freeze-thaw cycles. Because SAP_SP absorbs water in fresh mixtures, additional water was added to the mixture considering the water absorbency of the SAP_SP. It was found that the used SAP_SPs prematurely release their stored water so the effective water-to-cement ratio was increased when a larger amount of SAP_SP was used. The higher effective water-to-cement ratio caused more internal damage and salt scaling due to the weaker cementitious matrix. In addition, mortar samples with a high SAP_SP content show a larger absorption of capillary water than the reference sample. The result can be interpreted by an observation that SAP_SP in air voids absorbs water and expands to relatively large capillary pores or neighbor air voids during the capillary water absorption process.
Product Description
Millipore
PTFE Membrane Filter, 0.45 μm Pore Size, Omnipore, filter diam. 47 mm, hydrophilic
Millipore
Stainless Steel Pressure Filter Holder, 47 mm, 340 mL, Stainless Steel Pressure Filter Holder, 47mm, 340mL