MilliporeSigma
  • Nuclear prelamin a recognition factor and iron dysregulation in multiple sclerosis.
Nuclear prelamin a recognition factor and iron dysregulation in multiple sclerosis.
Metabolic brain disease (2019-12-12)
Di Ding, Anddre Osmar Valdivia, Sanjoy K Bhattacharya
ABSTRACT
Dysregulation of iron metabolism and aberrant iron deposition has been associated with multiple sclerosis. However, the factors that contribute to this pathological state remain to be understood. In this study, human multiple sclerosis and mice brain samples were analyzed through mass spectrometry as well as histological and immunoblot techniques, which demonstrated that iron deposition is associated with increased levels of nuclear prelamin A recognition factor (NARF). NARF is a protein associated with the mitochondria which has also been linked to mitochondrial defects in multiple sclerosis. We report NARF to be associated in multiple sclerosis pathology and aberrant iron deposition.
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
Iron Stain