MilliporeSigma
  • One bout of neonatal inflammation impairs adult respiratory motor plasticity in male and female rats.
One bout of neonatal inflammation impairs adult respiratory motor plasticity in male and female rats.
eLife (2019-03-23)
Austin D Hocker, Sarah A Beyeler, Alyssa N Gardner, Stephen M Johnson, Jyoti J Watters, Adrianne G Huxtable
ABSTRACT
Neonatal inflammation is common and has lasting consequences for adult health. We investigated the lasting effects of a single bout of neonatal inflammation on adult respiratory control in the form of respiratory motor plasticity induced by acute intermittent hypoxia, which likely compensates and stabilizes breathing during injury or disease and has significant therapeutic potential. Lipopolysaccharide-induced inflammation at postnatal day four induced lasting impairments in two distinct pathways to adult respiratory plasticity in male and female rats. Despite a lack of adult pro-inflammatory gene expression or alterations in glial morphology, one mechanistic pathway to plasticity was restored by acute, adult anti-inflammatory treatment, suggesting ongoing inflammatory signaling after neonatal inflammation. An alternative pathway to plasticity was not restored by anti-inflammatory treatment, but was evoked by exogenous adenosine receptor agonism, suggesting upstream impairment, likely astrocytic-dependent. Thus, the respiratory control network is vulnerable to early-life inflammation, limiting respiratory compensation to adult disease or injury.
Sigma-Aldrich
Anti-Substance P Receptor Antibody, Chemicon®, from guinea pig
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
CGS-21680 hydrochloride hydrate, solid
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Sigma-Aldrich
(S)-(+)-Ketoprofen, 99%