MilliporeSigma
  • Characteristics of stem cells from human neuroblastoma cell lines and in tumors.
Characteristics of stem cells from human neuroblastoma cell lines and in tumors.
Neoplasia (New York, N.Y.) (2005-02-22)
Jeanette D Walton, David R Kattan, Sharon K Thomas, Barbara A Spengler, Hong-Fen Guo, June L Biedler, Nai-Kong V Cheung, Robert A Ross
ABSTRACT
Cellular heterogeneity is a hallmark of human neuroblastoma tumors and cell lines. Within a single neuroblastoma are cells from distinct neural crest lineages whose relative abundance is significant for prognosis. We postulate that a self-renewing multipotent tumor stem cell, which gives rise to diverse cell lineages, is the malignant progenitor of this cancer. To test this hypothesis, we have established 22 cloned, phenotypically homogeneous populations of the three major cell types from 17 neuroblastoma cell lines. In vitro, malignant neuroblastoma stem cells, termed I-type (intermediate type), have distinct morphologic, biochemical, differentiative, and tumorigenic properties. I-type cells express features of both neuroblastic (N) cells (scant cytoplasm, neuritic processes, neurofilaments, pseudoganglia, and granin and neurotransmitter enzyme expression) and substrate-adherent (S) cells (extensive cytoplasm and vimentin and CD44 expression). Moreover, they show bidirectional differentiation to either N or S cells when induced by specific agents. I-type cells are significantly more malignant than N- or S-type cells, with four- to five-fold greater plating efficiencies in soft agar and six-fold higher tumorigenicity in athymic mice. Differences in malignant potential are unrelated to N-myc amplification/overexpression or the ability to digest and migrate through the extracellular matrix. Immunocytochemical analyses of a small series of tumors reveal that frequency of cells coexpressing N and S cell markers correlates with poor prognosis. Thus, I-type stem cells may be instrumental in the genesis and growth of tumors in the patient. Their unique biology deserves attention and further investigation.
Sigma-Aldrich
Anti-Neurofilament 70 kDa Antibody, clone DA2, culture supernatant, clone DA2, Chemicon®
Sigma-Aldrich
Anti-Heat Shock Protein 70 Antibody, clone W27, clone W27, Chemicon®, from mouse
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Vimentin Antibody, serum, Chemicon®