MilliporeSigma
  • Pro-soft Val-boroPro: a strategy for enhancing in vivo performance of boronic acid inhibitors of serine proteases.
Pro-soft Val-boroPro: a strategy for enhancing in vivo performance of boronic acid inhibitors of serine proteases.
Journal of medicinal chemistry (2011-03-11)
Sarah E Poplawski, Jack H Lai, David G Sanford, James L Sudmeier, Wengen Wu, William W Bachovchin
ABSTRACT
Val-boroPro, 1, is a potent, but relatively nonspecific inhibitor of the prolyl peptidases. It has antihyperglycemic activity from inhibition of DPPIV but also striking anticancer activity and a toxicity for which the mechanisms are unknown. 1 cyclizes at physiological pH, which attenuates its inhibitory potency >100-fold, which is a "soft drug" effect. Here we show that this phenomenon can be exploited to create prodrugs with unique properties and potential for selective in vivo targeting. Enzyme-mediated release delivers 1 to the target in the active form at physiological pH; cyclization attenuates systemic pharmacological effects from subsequent diffusion. This "pro-soft" design is demonstrated with a construct activated by and targeted to DPPIV, including in vivo results showing improved antihyperglycemic activity and reduced toxicity relative to 1. Pro-soft derivatives of 1 can help to illuminate the mechanisms underlying the three biological activities, or to help localize 1 at a tumor and thereby lead to improved anticancer agents with reduced toxicity. The design concept can also be applied to a variety of other boronic acid inhibitors.
Sigma-Aldrich
Val-boroPro