MilliporeSigma
  • Exchange protein directly activated by cAMP (EPAC) promotes transcriptional activation of the decidual prolactin gene via CCAAT/enhancer-binding protein in human endometrial stromal cells.
Exchange protein directly activated by cAMP (EPAC) promotes transcriptional activation of the decidual prolactin gene via CCAAT/enhancer-binding protein in human endometrial stromal cells.
Reproduction, fertility, and development (2018-05-08)
Kazuya Kusama, Kazuhiro Tamura, Hanako Bai, Toshihiro Sakurai, Hirotaka Nishi, Keiichi Isaka, Kazuhiko Imakawa, Mikihiro Yoshie
ABSTRACT
Protein kinase A (PKA) signalling accompanies elevated intracellular cAMP levels during endometrial stromal cell (ESC) decidualisation. Exchange protein directly activated by cAMP (EPAC), an alternate mediator of cAMP signalling, promotes PKA analogue-induced decidualisation; however, the precise mechanism by which EPAC and PKA co-operatively stimulate decidualisation has not been characterised. To examine the role of CCAAT/enhancer-binding protein (C/EBP) in EPAC- and PKA-mediated decidualisation of primary human ESCs, a reporter plasmid containing the 332bp region upstream from the transcription initiation site of the decidual prolactin (dPRL) gene was generated and the promoter activity was evaluated using a luciferase assay. The dPRL promoter activity was increased by treatment of transfected ESCs with the PKA-selective cAMP analogue N6-phenyl-cAMP (Phe) and enhanced further by co-treatment with the EPAC-selective cAMP analogue 8-(4-chlorophenyltio)-2'-O-methyl cAMP (CPT). Treatment with forskolin, an adenylyl cyclase activator, had a similar effect on reporter activity. Site-directed mutagenesis of the C/EBPβ- and/or C/EBPδ-binding site in the dPRL promoter abolished Phe/CPT-mediated elevation of the reporter activity. EPAC2 knockdown markedly reduced Phe-stimulated C/EBPβ and C/EBPδ mRNA levels, as well as forkhead box O1 (FOXO1) protein levels. These results suggest that EPAC signalling enhances PKA-mediated dPRL expression in ESCs by acting on C/EBP response elements in the promoter region of the gene.
Sigma-Aldrich
MISSION® esiRNA, targeting human RAPGEF3
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human RAPGEF4