MilliporeSigma
  • Aortic carboxypeptidase-like protein, a WNT ligand, exacerbates nonalcoholic steatohepatitis.
Aortic carboxypeptidase-like protein, a WNT ligand, exacerbates nonalcoholic steatohepatitis.
The Journal of clinical investigation (2018-03-20)
Toshiaki Teratani, Kengo Tomita, Takahiro Suzuki, Hirotaka Furuhashi, Rie Irie, Makoto Nishikawa, Junji Yamamoto, Toshifumi Hibi, Soichiro Miura, Tohru Minamino, Yuichi Oike, Ryota Hokari, Takanori Kanai
ABSTRACT
Incidence of nonalcoholic steatohepatitis (NASH), which is considered a hepatic manifestation of metabolic syndrome, has been increasing worldwide with the rise in obesity; however, its pathological mechanism is poorly understood. Here, we demonstrate that the hepatic expression of aortic carboxypeptidase-like protein (ACLP), a glycosylated, secreted protein, increases in NASH in humans and mice. Furthermore, we elucidate that ACLP is a ligand, unrelated to WNT proteins, that activates the canonical WNT pathway and exacerbates NASH pathology. In the liver, ACLP is specifically expressed in hepatic stellate cells (HSCs). As fatty liver disease progresses, ACLP expression is enhanced via activation of STAT3 signaling by obesity-related factors in serum. ACLP specifically binds to frizzled-8 and low-density lipoprotein-related receptor 6 to form a ternary complex that activates canonical WNT signaling. Consequently, ACLP activates HSCs by inhibiting PPARγ signals. HSC-specific ACLP deficiency inhibits fibrosis progression in NASH by inhibiting canonical WNT signaling in HSCs. The present study elucidates the role of canonical WNT pathway activation by ACLP in NASH pathology, indicating that NASH can be treated by targeting ACLP-induced canonical WNT pathway activation in HSCs.
Sigma-Aldrich
MISSION® esiRNA, targeting human FZD6
Sigma-Aldrich
MISSION® esiRNA, targeting human LRP6
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)