Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU027961

Sigma-Aldrich

MISSION® esiRNA

targeting human LRP6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCATGCACCTGGTTCTACTTCAAGATGAGCTATCATGTGGAGAACCTCCAACATGTTCTCCTCAGCAGTTTACTTGTTTCACGGGGGAAATTGACTGTATCCCTGTGGCTTGGCGGTGCGATGGGTTTACTGAATGTGAAGACCACAGTGATGAACTCAATTGTCCTGTATGCTCAGAGTCCCAGTTCCAGTGTGCCAGTGGGCAGTGTATTGATGGTGCCCTCCGATGCAATGGAGATGCAAACTGCCAGGACAAATCAGATGAGAAGAACTGTGAAGTGCTTTGTTTAATTGATCAGTTCCGCTGTGCCAATGGTCAGTGCATTGGAAAGCACAAGAAGTGTGATCATAATGTGGATTGCAGTGACAAGTCAGATGAACTGGATTGTTATCCGACTGAAGAACCAGCACCACAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Luo et al.
Neuroscience bulletin, 36(10), 1171-1181 (2020-06-21)
Neuronal apoptosis is one of the essential mechanisms of early brain injury after subarachnoid hemorrhage (SAH). Recently, HLY78 has been shown to inhibit apoptosis in tumor cells and embryonic cells caused by carbon ion radiation through activation of the Wnt/β-catenin
Xu Luo et al.
Brain research bulletin, 162, 107-114 (2020-06-23)
Wnt/β-catenin signaling plays an essential role in blood-brain barrier (BBB) formation and maintenance under pathophysiological conditions. HLY78, a lycorine derivative, has been identified as a novel activator of Wnt/β-catenin signaling in vitro. However, the effects of HLY78 on the BBB
Han Zhou et al.
JCI insight, 5(3) (2020-02-14)
Vascular inflammation is present in many cardiovascular diseases, and exogenous glucocorticoids have traditionally been used as a therapy to suppress inflammation. However, recent data have shown that endogenous glucocorticoids, acting through the endothelial glucocorticoid receptor, act as negative regulators of
Lei Wang et al.
Bone research, 6, 22-22 (2018-07-25)
Low-density lipoprotein receptor-related protein 6 (LRP6) is a co-receptor for Wnt signaling and can be recruited by multiple growth factors/hormones to their receptors facilitating intracellular signaling activation. The ligands that bind directly to LRP6 have not been identified. Here, we
Yaqi Qiu et al.
Cell and tissue research, 383(3), 1077-1092 (2020-11-28)
Bile salt-dependent lipase (BSDL) within intestinal lumen can be endocytosed by enterocytes and support the intestinal barrier function. However, the epithelial-supporting effect of this protein has not been verified in a human cell line and neither the direct signaling pathway

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service