MilliporeSigma
  • Folate receptor-targeted lipid-albumin nanoparticles (F-LAN) for therapeutic delivery of an Akt1 antisense oligonucleotide.
Folate receptor-targeted lipid-albumin nanoparticles (F-LAN) for therapeutic delivery of an Akt1 antisense oligonucleotide.
Journal of drug targeting (2018-01-30)
Hong Li, Yang Liu, Lihua Chen, Qibing Liu, Shanshan Qi, Xinwei Cheng, Young B Lee, Chang-Ho Ahn, Deog Joong Kim, Robert J Lee
ABSTRACT
RX-0201 is an antisense oligonucleotide (ASO) against Akt1 currently in clinical trial for metastatic renal cancer. To improve the delivery of RX-0201 using folate receptor-targeted lipid-albumin nanoparticles (F-LAN). F-LAN were synthesized with the composition of DOTAP/soyPC/TPGS/folate-PEG-DSPE (25:70:4:1 m/m), a cationic human serum albumin-pentaethylenehexamine (HSA-PEHA) conjugate and RX-0201. The nanoparticles were evaluated in KB human carcinoma cells in vitro and in a KB murine xenograft tumour model in vivo for pharmacokinetics and antitumor activities. The F-LAN-RX-0201 had a mean particle size of 108.6 ± 5.8 nm, zeta potential of 10.5 ± 3.2 mV and ASO loading efficiency of 71.5 ± 4.5%. In KB cells, uptake and Akt1 inhibition by F-LAN-RX-0201 were greater than those of non-targeted LAN-RX-0201 and could be partially blocked by excess free folate. F-LAN-RX-0201 inhibited cell growth with an IC50 of 11.9 μM. In contrast, LAN-RX-0201 showed lower cytotoxicity with an IC50 of 32.0 μM. No significant cytotoxicity was observed with up to 250 µM of free RX-0201. Pharmacokinetic studies showed that F-LAN-RX-0201 had a longer terminal half-life than free RX-0201 (442 vs. 219 min). In a KB xenograft tumour model, F-LAN-RX-0201 exhibited greater tumour inhibition than LAN-RX-0201 at 16 mg/kg. Moreover, F-LAN-RX-0201 at 16 mg/kg showed comparable tumour inhibition compared to free RX-0201 at a much higher dose of 90 mg/kg. F-LAN-RX-0201 showed promise as a therapeutic agent for tumours with elevated folate-receptor expression.
Sigma-Aldrich
Pentaethylenehexamine, technical grade