MilliporeSigma
  • A rapid solution-based method for determining the affinity of heroin hapten-induced antibodies to heroin, its metabolites, and other opioids.
A rapid solution-based method for determining the affinity of heroin hapten-induced antibodies to heroin, its metabolites, and other opioids.
Analytical and bioanalytical chemistry (2018-04-21)
Oscar B Torres, Alexander J Duval, Agnieszka Sulima, Joshua F G Antoline, Arthur E Jacobson, Kenner C Rice, Carl R Alving, Gary R Matyas
ABSTRACT
We describe for the first time a method that utilizes microscale thermophoresis (MST) technology to determine polyclonal antibody affinities to small molecules. Using a novel type of heterologous MST, we have accurately measured a solution-based binding affinity of serum antibodies to heroin which was previously impossible with other currently available methods. Moreover, this mismatch approach (i.e., using a cross-reactive hapten tracer) has never been reported in the literature. When compared with equilibrium dialysis combined with ultra-performance liquid chromatography/tandem mass spectrometry (ED-UPLC/MS/MS), this novel MST method yields similar binding affinity values for polyclonal antibodies to the major heroin metabolites 6-AM and morphine. Additionally, we herein report the method of synthesis of this novel cross-reactive hapten, MorHap-acetamide-a useful analog for the study of heroin hapten-antibody interactions. Using heterologous MST, we were able to determine the affinities, down to nanomolar accuracies, of polyclonal antibodies to various abused opioids. While optimizing this method, we further discovered that heroin is protected from serum esterase degradation by the presence of these antibodies in a concentration-dependent manner. Lastly, using affinity data for a number of structurally different opioids, we were able to dissect the moieties that are crucial to antibody binding. The novel MST method that is presented herein can be extended to the analysis of any ligand that is prone to degradation and can be applied not only to the development of vaccines to substances of abuse but also to the analysis of small molecule/protein interactions in the presence of serum. Graphical abstract Strategy for the determination of hapten-induced antibody affinities using Microscale thermophoresis.
Sigma-Aldrich
Bis(4-nitrophenyl) phosphate, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
N-Methylpiperidine, 99%
Pricing and availability is not currently available.