Skip to Content
MilliporeSigma
PYREXPLUS® Coated Round Media Storage Bottles capacity 250 mL

PYREXPLUS® Coated Round Media Storage Bottles

PYREXPLUS® Coated Round Media Storage Bottles capacity 250 mL
Synonym(s):
Corning® 61626

Products

Product Category
Manufacturer
Capacity (mL)
Diameter (mm)
Height (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size