Skip to Content
MilliporeSigma
Platinum(IV) chloride (57.5% Pt) anhydrous, for synthesis

Platinum(IV) chloride

Platinum(IV) chloride (57.5% Pt) anhydrous, for synthesis
Synonym(s):
Platinum tetrachloride, Platinum(IV) chloride (57.5% Pt)
Linear Formula:
PtCl4
CAS Number:
Molecular Weight:
336.89

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing