Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.07347

Sigma-Aldrich

Platinum(IV) chloride

57.5% Pt, anhydrous, for synthesis

Synonym(s):

Platinum(IV) chloride (57.5% Pt), Platinum tetrachloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$83.20
1 G
$309.00
5 G
$1,200.00

$83.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$83.20
1 G
$309.00
5 G
$1,200.00

About This Item

Empirical Formula (Hill Notation):
Cl4Pt
CAS Number:
Molecular Weight:
336.89
MDL number:
UNSPSC Code:
12352302
EC Index Number:
236-645-1
NACRES:
NA.22
grade:
for synthesis
synthesis grade
form:
powder
solubility:
587 g/L

$83.20


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Platinum(IV) chloride (57.5% Pt), anhydrous, for synthesis

grade

for synthesis
synthesis grade

Quality Level

form

powder

potency

276 mg/kg LD50, oral (Rat)

pH

<1 (20 °C in H2O)

solubility

587 g/L

density

4.3 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/4Cl.Pt

InChI key

YTQAIHLQJINUGP-UHFFFAOYSA-N

Analysis Note

Assay (ex Pt): ≥ 99.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service