
Phosphate buffer solution

Synonym(s):
Phosphate Buffer
Products
Product Category
Brand
Feature
pH Value
Physical Form
Product Line
Agency
Capacity (mL)
Quality Segment
Closure Type
Application
Package Size
Sterility
Sterilization Method
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing