Skip to Content
MilliporeSigma
Luperox® DDM-9, 2-Butanone peroxide solution ~35 wt. % in 2,2,4-trimethyl-1,3-pentanediol diisobutyrate

Luperox® DDM-9, 2-Butanone peroxide solution

Luperox® DDM-9, 2-Butanone peroxide solution ~35 wt. % in 2,2,4-trimethyl-1,3-pentanediol diisobutyrate
Synonym(s):
2-Butanone peroxide solution, Methyl ethyl ketone peroxide solution
Linear Formula:
C2H5C(OOH)(CH3)OOC(OOH)(CH3)C2H5
CAS Number:
Molecular Weight:
210.22

Products

Product Category
Brand
Formula Weight
Chemical Composition
Quality Segment
Markush Class
Markush Group
Reagent Type