Skip to Content
CA
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hexyl butyrate
Synonym(s):
Hexyl butyrate
Linear Formula:
CH
3
CH
2
CH
2
CO
2
(CH
2
)
5
CH
3
CAS Number:
2639-63-6
Molecular Weight:
172.26
EC Number:
220-136-6
Products
Product Category
aroma chemicals (2)
Brand
Sigma-Aldrich (3)
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Food Grade (2)
Halal (2)
Fragrance grade (1)
Kosher (1)
Natural (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
W256803
Hexyl butyrate,
≥98%, FG
Expand
View Pricing
8.14992
Hexyl butyrate,
for synthesis
Expand
View Pricing
W256811
Hexyl butyrate,
natural, ≥98%, FG
Expand
View Pricing