Skip to Content
MilliporeSigma
Ethylenediaminetetraacetic acid BioUltra, ≥99.0% (KT)

Ethylenediaminetetraacetic acid

Ethylenediaminetetraacetic acid BioUltra, ≥99.0% (KT)
Synonym(s):
(Ethylenedinitrilo)tetraacetic acid, EDTA, Edathamil, Ethylenedinitrilotetraacetic acid
Linear Formula:
(HO2CCH2)2NCH2CH2N(CH2CO2H)2
CAS Number:
Molecular Weight:
292.24
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Reagent Type
Special Grade
Technique