Skip to Content
MilliporeSigma
Brodifacoum PESTANAL®, analytical standard

Brodifacoum

Brodifacoum PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C31H23BrO3
CAS Number:
Molecular Weight:
523.42
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing