Skip to Content
MilliporeSigma
Anti-MACC1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-MACC1 antibody produced in rabbit

Anti-MACC1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-Metastasis associated in colon cancer 1, Anti-SH3 domain-containing protein 7a5, Anti-7A5, Anti-CAGE, Anti-CT26, Anti-PHB1, Anti-SH3BP4L, Anti-metastasis associated in colon cancer 1

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Species Reactivity
Technique