Skip to Content
MilliporeSigma
4,4,4,4′,4′,4′-Hexafluoro-DL-valine 97%

4,4,4,4′,4′,4′-Hexafluoro-DL-valine

4,4,4,4′,4′,4′-Hexafluoro-DL-valine 97%
Synonym(s):
2-Amino-4,4,4-trifluoro-3-(trifluoromethyl)butyric acid, 4,4,4,5,5,5-Hexafluoro-DL-valine
Linear Formula:
(CF3)2CHCH(NH2)COOH
CAS Number:
Molecular Weight:
225.09
EC Number:

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing