Skip to Content
MilliporeSigma
1-(Trimethylsilyl)imidazole for GC derivatization, LiChropur™

1-(Trimethylsilyl)imidazole

1-(Trimethylsilyl)imidazole for GC derivatization, LiChropur™
Synonym(s):
N-Trimethylsilylimidazole, TMSI, TSIM
Empirical Formula (Hill Notation):
C6H12N2Si
CAS Number:
Molecular Weight:
140.26
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Technique
Showing 1-3 of 3