Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900929

Sigma-Aldrich

Penicillin-Streptomycin

suitable for cell culture, Vetec, BioReagent

Synonym(s):

Penicillin-Streptomycin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

Product Name

Penicillin-Streptomycin, Vetec, reagent grade, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, sterile, BioReagent, suitable for cell culture

grade

reagent grade

Quality Level

sterility

sterile

product line

BioReagent
Vetec

technique(s)

cell culture | mammalian: suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

−20°C

SMILES string

CC1(C)S[C@@H]2[C@H](NC(=O)Cc3ccccc3)C(=O)N2[C@H]1C(O)=O.CNC[C@H]4[C@H](O)[C@@H](O)[C@H](CCO)O[C@H]4O[C@H]5[C@@H](O[C@@H](C)C5(O)C=O)O[C@H]6[C@H](O)[C@@H](O)[C@H](NC(N)=N)[C@@H](O)[C@@H]6NC(N)=N

InChI

1S/C23H43N7O12.C16H18N2O4S/c1-7-23(38,6-32)18(42-19-8(5-28-2)12(33)13(34)9(40-19)3-4-31)20(39-7)41-17-11(30-22(26)27)14(35)10(29-21(24)25)15(36)16(17)37;1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9/h6-20,28,31,33-38H,3-5H2,1-2H3,(H4,24,25,29)(H4,26,27,30);3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22)/t7-,8-,9-,10+,11-,12-,13-,14+,15-,16+,17+,18-,19-,20-,23+;11-,12+,14-/m01/s1

InChI key

KMOOCZWLFBSQCW-XFOLRDBHSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fengliang Xu et al.
Biochemical and biophysical research communications, 516(2), 526-532 (2019-06-25)
The hepatocellular carcinoma (HCC) is a common and highly aggressive malignancy especially in China. Accumulating data have shown a critical role of long non-coding RNAs (lncRNAs) during cancer progression. However, the function of lncRNA TSLD8 remains elusive. By lncRNA profiling
Tingting Zhao et al.
Naunyn-Schmiedeberg's archives of pharmacology, 393(6), 1013-1024 (2019-12-12)
Ischemia-reperfusion (I/R) injury is a kind of the tissue damage caused by an abrupt re-supplying tissue with blood after a period of ischemia or hypoxia. It contributes to a wide range of pathological processes including kidney injury, circulatory arrest, and
Lu Li et al.
Biochemical and biophysical research communications, 492(3), 289-294 (2017-09-03)
Twinfilin1 (TWF1) is an actin monomer-binding protein, which biological function has not yet been fully uncovered. In our previous study, we found by mass spectrometry analysis that TWF1 might be one of the major proteins responsible for milk bio-synthesis and
Kai-Liang Zhang et al.
OncoTargets and therapy, 12, 6515-6523 (2019-10-17)
The efficacy of traditional therapies for oral carcinoma (OC) is limited. Oncolytic adenovirus, a novel strategy of cancer therapy, shows potential use in OC treatment. However, its clinical application is limited by pre-existing neutralizing antibodies. Thus, this study aimed to
Lunzhi Yuan et al.
Emerging microbes & infections, 7(1), 144-144 (2018-08-12)
The human liver chimeric mouse with primary human hepatocytes (PHHs) engraftment has been demonstrated to be a useful animal model to study hepatitis B virus (HBV) pathogenesis and evaluate anti-HBV drugs. However, the disadvantages of using PHHs include the inability

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service