Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900868

Sigma-Aldrich

Guanosine 5′-triphosphate sodium salt hydrate

Vetec, reagent grade, ≥95%, powder

Synonym(s):

GTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N5O14P3 · xNa+
CAS Number:
Molecular Weight:
523.18 (free acid basis)
Beilstein:
3585677
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C10H16N5O14P3.3Na/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(27-9)1-26-31(22,23)29-32(24,25)28-30(19,20)21;;;/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H,24,25)(H2,19,20,21)(H3,11,13,14,18);;;/q;3*+1/p-3/t3-,5-,6-,9-;;;/m1.../s1

InChI key

KZRMTEVIDYXWQW-CYCLDIHTSA-K

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

GTP functions as a carrier of phosphates and pyrophosphates involved in channeling chemical energy into specific biosynthetic pathways. GTP activates the signal transducing G proteins which are involved in various cellular processes including proliferation, differentiation, and activation of several intracellular kinase cascades. Proliferation and apoptosis are regulated in part by the hydrolysis of GTP by small GTPases Ras and Rho. Another type of small GTPase, Rab, plays a role in the docking and fusion of vesicles and may also be involved in vesicle formation. In addition to its role in signal transduction, GTP also serves as an energy-rich precursor of mononucleotide units in the enzymatic biosynthesis of DNA and RNA.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A Cooper et al.
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 184(7), 865-876 (2014-08-28)
Marine teleost fish secrete bicarbonate (HCO3 (-)) into the intestine to aid osmoregulation and limit Ca(2+) uptake by carbonate precipitation. Intestinal HCO3 (-) secretion is associated with an equimolar transport of protons (H(+)) into the blood, both being proportional to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service