Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900858

Sigma-Aldrich

TRIS glycine SDS buffer solution

Vetec, reagent grade, 10 ×, solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319

grade

reagent grade

product line

Vetec

form

solution

concentration

10 ×

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service