Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900854

RIPA Buffer

Vetec, reagent grade,

protease free

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700

grade

reagent grade

product line

Vetec

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

RIPA Buffer enables efficient cell lysis and protein solubilization while avoiding protein degradation and interference with the proteins′ immunoreactivity and biological activity. RIPA Buffer also results in low background in immunoprecipitation and molecular pull-down assays. Compatible with EZview Affinity Gels.

Legal Information

EZview is a trademark of Sigma-Aldrich Co. LLC
Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Cui et al.
Experimental physiology, 103(12), 1618-1632 (2018-09-12)
What is the central question of this study? What is the role of hypothalamic DNA methylation in the development of polycystic ovary syndrome (PCOS) and the response to electro-acupuncture treatment. What is the main finding and its importance? Global DNA
Qingxia Gao et al.
Journal of virology, 95(2) (2020-10-23)
The viral ribonucleoprotein (vRNP) of the influenza A virus (IAV) is responsible for the viral RNA transcription and replication in the nucleus, and its functions rely on host factors. Previous studies have indicated that eukaryotic translation elongation factor 1 delta
Cha Yang et al.
Journal of virology, 93(21) (2019-08-16)
The innate immune response is vital for host defense and must be tightly controlled, but the mechanisms responsible for its negative regulation are not fully understood. The cell growth-regulating nucleolar protein LYAR was found to promote replication of multiple viruses
Weizhen Liu et al.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association, 23(1), 39-51 (2019-06-15)
Activating mutation of KIT or PDGFRA is the primary molecular mechanism for gastrointestinal stromal tumors (GISTs). Although imatinib has a revolutionary effect on GIST therapeutics, the benefits are not durable. Increasing reports have demonstrated that cell cycle checkpoint plays critical
Lei Shi et al.
The Journal of eukaryotic microbiology, 67(5), 521-531 (2020-05-06)
Intraflagellar transport (IFT) represents a bidirectional dynamic process that carries cargo essential for cilia building and the maintenance of ciliary function, which is important for the locomotion of single cells, intracellular and intercellular signalling transduction. Accumulated evidence has revealed that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service