Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900801

Sigma-Aldrich

Sodium carbonate

Vetec, reagent grade, ≥99.5%

Synonym(s):

Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
EC Number:
MDL number:
UNSPSC Code:
12352302
Assay:
≥99.5%
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

≥99.5%

form

powder

pH

12 (25 °C, 106 g/L)

mp

851 °C (lit.)

cation traces

heavy metals (as Pb): ≤10 ppm

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo Isanta et al.
Water research, 80, 149-158 (2015-05-23)
Partial nitritation for a low-strength wastewater at low temperature was stably achieved in an aerobic granular reactor. A bench-scale granular sludge bioreactor was operated in continuous mode treating an influent of 70 mg N-NH4(+) L(-1) to mimic pretreated municipal nitrogenous
Kun-Hui Lu et al.
Journal of leukocyte biology, 98(3), 301-311 (2015-05-23)
Understanding the molecular mechanisms regulating T cell reactivity is required for successful reprogramming of immune responses in medical conditions, characterized by dysfunctions of the immune system. Nck proteins are cytoplasmic adaptors mediating diverse cellular functions, including TCR signaling. By enhancing
I Irwansyah et al.
Advanced materials (Deerfield Beach, Fla.), 27(4), 648-654 (2014-12-03)
Antimicrobial hydrogels are prepared based on the co-assembly of commercial Fmoc-phenylalanine and Fmoc-leucine, which act as the hydrogelator and antimicrobial building block, respectively. This co-assembled antimicrobial hydrogel is demonstrated to exhibit selective bactericidal activity for gram-positive bacteria while being biocompatible
Jia Bao et al.
Environmental science and pollution research international, 21(12), 7650-7655 (2014-03-14)
Severe perfluoroalkyl acid (PFAA) contamination resulting from the fast-growing semiconductor, electrochemical, and optoelectronic industries has been determined in the river water in the vicinity of the Taipei area, Taiwan, during recent years. However, little is known about body burdens of
Yukiyo Sato et al.
PloS one, 9(6), e99041-e99041 (2014-06-11)
The accumulation of RCY1 protein, which is encoded by RESISTANCE TO CMV(Y) (RCY1), a CC-NB-LRR class R-gene, is tightly correlated with the strength of the resistance to a yellow strain of Cucumber mosaic virus [CMV(Y)] in Arabidopsis thaliana. In order

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service