Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900753

Sigma-Aldrich

Triethyl citrate

Vetec, reagent grade, 98%

Synonym(s):

Ethyl citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOC2H5)(CH2COOC2H5)2
CAS Number:
Molecular Weight:
276.28
Beilstein:
1801199
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

9.7 (vs air)

vapor pressure

1 mmHg ( 107 °C)

product line

Vetec

Assay

98%

refractive index

n20/D 1.442 (lit.)

bp

235 °C/150 mmHg (lit.)

density

1.14 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC

InChI

1S/C12H20O7/c1-4-17-9(13)7-12(16,11(15)19-6-3)8-10(14)18-5-2/h16H,4-8H2,1-3H3

InChI key

DOOTYTYQINUNNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ravikumar R Patel et al.
Acta pharmaceutica (Zagreb, Croatia), 61(1), 73-82 (2011-03-17)
In situ forming intragastric controlled-release formulation is a new technology in the field of oral controlled-release delivery systems. The objective of this study was to develop formulations that can control drug release up to 24 hours. In addition, a combination
T Quinten et al.
AAPS PharmSciTech, 13(4), 1197-1211 (2012-09-12)
Sustained-release matrix tablets based on Eudragit RL and RS were manufactured by injection moulding. The influence of process temperature; matrix composition; drug load, plasticizer level; and salt form of metoprolol: tartrate (MPT), fumarate (MPF) and succinate (MPS) on ease of
Shital M Maru et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 44(4), 471-478 (2011-09-20)
The influence of antiretroviral drugs and plasticizers on the rheological and thermal characteristics of ethyl cellulose formulations intended for hot melt extrusion has been investigated. Antiretroviral drugs used were zidovudine and lamivudine, whilst plasticizers included triethylcitrate (TEC) and polyethylene glycol
H M Fadda et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 76(3), 493-497 (2010-08-31)
Glass transition temperature (T(g)) measurements of polymers are conventionally conducted in the dry state with little attention to the environment they are designed to work in. Our aim was to develop the novel use of dynamic mechanical analysis (DMA) to
Claire Gendre et al.
International journal of pharmaceutics, 421(2), 237-243 (2011-10-11)
The aim of this work was to carry out real-time near infrared (NIR) predictions of drug release from sustained release coated tablets and to determine end point of coating operation. In-line measurements were ensured by implementation of a NIR probe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service