Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900731

Sigma-Aldrich

Dimethyl fumarate

Vetec, reagent grade, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH=CHCOOCH3
CAS Number:
Molecular Weight:
144.13
Beilstein:
774590
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

97%

bp

192-193 °C (lit.)

mp

102-106 °C (lit.)

SMILES string

[H]\C(=C(\[H])C(=O)OC)C(=O)OC

InChI

1S/C6H8O4/c1-9-5(7)3-4-6(8)10-2/h3-4H,1-2H3/b4-3+

InChI key

LDCRTTXIJACKKU-ONEGZZNKSA-N

Gene Information

human ... KEAP1(9817)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Meissner et al.
Journal der Deutschen Dermatologischen Gesellschaft = Journal of the German Society of Dermatology : JDDG, 10(11), 793-801 (2012-08-18)
Fumaric acid esters have been used successfully in the therapy of psoriasis vulgaris since 1959. In the last 17 years, many of the underlying mechanisms of anti-psoriatic action, such as a Th1/Th2 shift, a suppression of important leukocyte adhesion molecules
Ralf A Linker et al.
Current neurology and neuroscience reports, 13(11), 394-394 (2013-09-26)
Dimethyl fumarate is an orally available treatment option for relapsing-remitting multiple sclerosis (MS) in a new formulation with improved gastroenteric coating. The mode of action comprises immunomodulatory effects and an activation of nuclear (erythroid-derived 2) related factor mediated antioxidative response
Frieder Kees
Expert opinion on pharmacotherapy, 14(11), 1559-1567 (2013-05-24)
Dimethyl fumarate (DMF) has been used as fungicide, but oral DMF activates anti-inflammatory and anti-oxidative pathways that are beneficial in the treatment of psoriasis. BG-12, a specific formulation of DMF, has been approved very recently for the treatment of relapsing-remitting
Take-Aki Mitsudo et al.
Chemical record (New York, N.Y.), 6(3), 107-116 (2006-06-30)
The chemistry of a novel zerovalent Ru complex, Ru(eta6-cot)(eta2-dmfm)2 (1) (cot=1,3,5-cyclooctatriene; dmfm=dimethyl fumarate), is reviewed with a focus on its reactivity toward phosphines, amines, and H2O, as well as arenes and p-quinones. A variety of novel zerovalent Ru complexes were
Masanobu Tsubaki et al.
European journal of cancer (Oxford, England : 1990), 49(17), 3708-3717 (2013-08-13)
Several autocrine soluble factors, including macrophage inflammatory protein-1α and tumour necrosis factor-alpha (TNF-α), promote the survival and growth of multiple myeloma (MM) cells. We hypothesised that inhibition of the TNF-α autocrine loop may enhance the cytotoxic effect of anticancer drugs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service