Skip to Content
MilliporeSigma
All Photos(2)

Documents

V900705

Sigma-Aldrich

Trimethylamine hydrochloride

Vetec, reagent grade, 98%

Synonym(s):

Trimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N · HCl
CAS Number:
Molecular Weight:
95.57
Beilstein:
3905063
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

mp

283-284 °C (dec.) (lit.)

SMILES string

Cl[H].CN(C)C

InChI

1S/C3H9N.ClH/c1-4(2)3;/h1-3H3;1H

InChI key

SZYJELPVAFJOGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jong Hyun Lim et al.
Biosensors & bioelectronics, 39(1), 244-249 (2012-08-21)
We herein report a peptide receptor-based bioelectronic nose (PRBN) that can determine the quality of seafood in real-time through measuring the amount of trimethylamine (TMA) generated from spoiled seafood. The PRBN was developed using single walled-carbon nanotube field-effect transistors (SWNT-FETs)
Smaranda Craciun et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(52), 21307-21312 (2012-11-16)
Choline and trimethylamine (TMA) are small molecules that play central roles in biological processes throughout all kingdoms of life. These ubiquitous metabolites are linked through a single biochemical transformation, the conversion of choline to TMA by anaerobic microorganisms. This metabolic
Qian Li et al.
Current biology : CB, 23(1), 11-20 (2012-11-28)
Rodents use olfactory cues for species-specific behaviors. For example, mice emit odors to attract mates of the same species, but not competitors of closely related species. This implies rapid evolution of olfactory signaling, although odors and chemosensory receptors involved are
S R Irish et al.
Journal of medical entomology, 50(3), 619-623 (2013-06-28)
Gravid traps are useful tools for monitoring vector-borne pathogens in mosquitoes, particularly for those pathogens transmitted by Culex quinquefasciatus Say. One of the primary challenges in the use of gravid traps is the necessity of the inclusion of an oviposition
Ivonne Wallrabenstein et al.
PloS one, 8(2), e54950-e54950 (2013-02-09)
In addition to the canonical olfactory receptors, TAARs were currently suggested to be a second class of chemosensory receptors in the olfactory epithelium of vertebrates. In contrast to several deorphanized murine TAARs, agonists for the intact human TAAR genes 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service