Skip to Content
MilliporeSigma
All Photos(2)

Documents

V900685

Sigma-Aldrich

3-Aminobutanoic acid

Vetec, reagent grade, 97%

Synonym(s):

(±)-3-Aminobutyric acid, DL-3-Aminobutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)CH2COOH
CAS Number:
Molecular Weight:
103.12
Beilstein:
1720563
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

97%

form

powder or chunks

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

189 °C (dec.) (lit.)

SMILES string

CC(N)CC(O)=O

InChI

1S/C4H9NO2/c1-3(5)2-4(6)7/h3H,2,5H2,1H3,(H,6,7)

InChI key

OQEBBZSWEGYTPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William Kay et al.
The FEBS journal, 273(13), 3002-3013 (2006-06-09)
Lipopolysaccharide (LPS) and beta-glucan from Francisella victoria, a fish pathogen and close relative of highly virulent mammal pathogen Francisella tularensis, have been analyzed using chemical and spectroscopy methods. The polysaccharide part of the LPS was found to contain a nonrepetitive
Zahed Hossain et al.
Journal of proteomics, 75(13), 4151-4164 (2012-06-02)
The present study highlights the protective role of β-aminobutyric acid (BABA) in alleviating cadmium (Cd) stress in soybean. Proteomic analyses revealed that out of 66 differentially abundant protein spots in response to Cd challenge, 17 were common in the leaves
Juhan Kim et al.
Applied and environmental microbiology, 73(6), 1772-1782 (2007-01-30)
A novel beta-transaminase gene was cloned from Mesorhizobium sp. strain LUK. By using N-terminal sequence and an internal protein sequence, a digoxigenin-labeled probe was made for nonradioactive hybridization, and a 2.5-kb gene fragment was obtained by colony hybridization of a
Chia-Hong Tsai et al.
The Plant journal : for cell and molecular biology, 65(3), 469-479 (2011-01-27)
The priming agent β-aminobutyric acid (BABA) is known to enhance Arabidopsis resistance to the bacterial pathogen Pseudomonas syringae pv. tomato (Pst) DC3000 by potentiating salicylic acid (SA) defence signalling, notably PR1 expression. The molecular mechanisms underlying this phenomenon remain unknown.
Eleonora Barilli et al.
Journal of proteomics, 75(17), 5189-5205 (2012-07-18)
Systemic acquired resistance (SAR) to Uromyces pisi in pea was studied by using a proteomic approach. Two-dimensional electrophoresis (2-DE) was used in order to compare the leaf proteome of two pea genotypes displaying different phenotypes (susceptible and partial resistance to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service