Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900683

Sigma-Aldrich

Citral

Vetec, reagent grade, 94%

Synonym(s):

3,7-Dimethyl-2,6-octadienal, Geranial and neral mixture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCHO
CAS Number:
Molecular Weight:
152.23
Beilstein:
1721871
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

5 (vs air)

vapor pressure

0.2 mmHg ( 200 °C)

product line

Vetec

Assay

94%

refractive index

n20/D 1.488 (lit.)

bp

229 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)C=C(C)CC\C=C(\C)C

InChI

1S/C10H16O/c1-9(2)5-4-6-10(3)7-8-11/h5,7-8H,4,6H2,1-3H3

InChI key

WTEVQBCEXWBHNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jon Lalko et al.
Regulatory toxicology and pharmacology : RTP, 52(1), 62-73 (2008-03-21)
Citral [CAS# 5392-40-5; EINECS# 226-394-6; RIFM # 116; cis- and trans-3,7-dimethyl-2,6-Octadienal] is an important fragrance ingredient appreciated for its powerful lemon-aroma. It is widely used in fragrance formulations and incorporated into numerous consumer products. A comprehensive review of the dermal
M Machado et al.
Experimental parasitology, 130(3), 223-231 (2012-01-10)
In order to contribute for the search of new drugs for leishmaniasis, we study the susceptibility of Leishmania infantum, Leishmania tropica and Leishmania major to Cymbopogon citratus essential oil and major compounds, mrycene and citral. C. citratus and citral were
Masrat Maswal et al.
Food chemistry, 138(4), 2356-2364 (2013-03-19)
Citral is a flavour component widely used in food and cosmetic industries, but is chemically unstable and degrades over time in aqueous solutions due to acid-catalysed and oxidative reactions leading to loss of desirable flavour. The present study reveals the
Francesca Patrignani et al.
International journal of food microbiology, 160(3), 273-281 (2013-01-08)
High pressure homogenization (HPH) technique is able to significantly reduce spoilage microbiota in fruit juice. On the other hand, aroma compounds and essential oils can have a key role in the microbial stability of these products. For this reason, the
Lawrence E Marks et al.
Physiology & behavior, 105(2), 443-450 (2011-09-21)
Stimulus context affects judgments of intensity of both gustatory and olfactory flavors, and the contextual effects are modality-specific. Does context also exert separate effects on the gustatory and olfactory components of flavor mixtures? To answer this question, in each of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service