V900671
N-Hydroxyphthalimide
Vetec™, reagent grade, 97%
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C8H5NO3
CAS Number:
Molecular Weight:
163.13
Beilstein:
131208
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
grade
reagent grade
product line
Vetec™
Assay
97%
reaction suitability
reaction type: Addition Reactions
mp
233 °C (dec.) (lit.)
SMILES string
ON1C(=O)c2ccccc2C1=O
InChI
1S/C8H5NO3/c10-7-5-3-1-2-4-6(5)8(11)9(7)12/h1-4,12H
InChI key
CFMZSMGAMPBRBE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
[Hydroxyphthalimide induced medicamentosa-like dermatitis in five patients].
Jian-fang Zou et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 24(10), 625-625 (2006-11-14)
S Coseri et al.
Biomacromolecules, 10(8), 2294-2299 (2009-09-03)
The oxidation reaction of regenerated cellulose fibers mediated by N-hydroxyphthalimide (NHPI) and various cocatalysts at room temperature for different time intervals and various amounts of low concentration sodium hypochlorite solution has been investigated to produce oxidized cellulose (OC), a biocompatible
V Krishnakumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(1), 110-115 (2008-01-26)
Single crystals of N-hydroxyphthalimide (NHPH) were obtained from saturated aqueous solutions of methanol and acetonitrile by slow cooling method. The grown crystals were bright and transparent. From the crystal structure analysis it can be inferred that the crystal belongs to
Contact allergy to N-hydroxyphthalimide.
S Fregert et al.
Contact dermatitis, 9(1), 84-85 (1983-01-01)
James M Takacs et al.
Organic letters, 5(20), 3595-3598 (2003-09-26)
[reaction: see text] Screening combinations of five catalyst precursors with 13 phosphorus ligands identified cyclization catalysts that favor carbocyclization-trapping with N-hydroxyphthalimide over a competing cycloisomerization mode.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service