Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900633

Sigma-Aldrich

Sodium 1-dodecanesulfonate

Vetec, reagent grade

Synonym(s):

1-Dodecanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11SO3Na
CAS Number:
Molecular Weight:
272.38
Beilstein:
3919536
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

mp

>300 °C (lit.)

SMILES string

[Na+]

InChI

1S/C12H26O3S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DAJSVUQLFFJUSX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yixiao Xu et al.
The American journal of Chinese medicine, 42(4), 799-816 (2014-07-10)
Pulmonary arterial hypertension (PAH) is a disease of the small pulmonary arteries characterized by increased vascular resistance. Pulmonary vasoconstriction has been proven to play a pivotal role in PAH. We have previously hypothesized that Panax notoginseng saponins (PNS) might attenuate
Manuel Mora et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(3), 989-995 (2011-01-14)
An Mg/Al layered double hydroxide (LDH) containing carbonate ion in its interlayer region was examined by medium infrared (MIR) and near infrared reflectance spectroscopy (NIRS). The MIR and NIR spectroscopy techniques was also used to study two organo-hybrid LDHs containing
Toshihide Tsukatani et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 22(2), 199-200 (2006-03-04)
The tetraoctylammonium cation forms water-immiscible room temperature ionic liquids with dodecylsulfate and dodecylbenzenesulfonate anions. The ionic liquids are halogen-free and can be considered environmentally friendly solvents. At 25 degrees C, the solubilities of water in tetraoctylammonium dodecylsulfate and tetraoctylammonium dodecylbenzenesulfonate
D Zielinska et al.
Chemosphere, 40(3), 327-330 (2000-02-09)
The objective of this study was to determine the interaction of modified bilayer lipid membranes (BLMs) with lead and tin organoderivatives. The relative depolarization, of the lipid membrane, caused by organo metals, was used to estimate their activity. Dodecyltrimethylammonium bromide
Jamil Rima et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 74(1), 10-15 (2009-07-14)
The solubilization of pyrene in aqueous solution of beta-cyclodextrin (beta-CD) or its derivatives such as beta-CD-hexanoyl, beta-CD-benzoyl and beta-CD-dodecylsulfonate was investigated by spectrophotometry. Linear and non-linear regression methods were used to estimate the association constants (K(1)). A 1:1 stoichiometric ratio

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service