Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900613

Sigma-Aldrich

D-(−)-Tartaric acid

Vetec, reagent grade, 99%

Synonym(s):

(2S,3S)-(−)-Tartaric acid, D-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725145
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

reagent grade

description

synthetic

product line

Vetec

Assay

99%

mp

172-174 °C (lit.)

SMILES string

O[C@@H]([C@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m0/s1

InChI key

FEWJPZIEWOKRBE-LWMBPPNESA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Unnatural isomer

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmen-Alice Teacă et al.
Carbohydrate polymers, 93(1), 307-315 (2013-03-08)
The present paper describes the preparation and characterization of polysaccharides-based bio-composite films obtained by the incorporation of 10, 20 and 30 wt% birch cellulose (BC) within a glycerol plasticized matrix constituted by the corn starch (S) and chemical modified starch
Anne Toboldt et al.
Applied and environmental microbiology, 78(20), 7347-7357 (2012-08-14)
In this study, the population structure, incidence, and potential sources of human infection caused by the d-tartrate-fermenting variant of Salmonella enterica serovar Paratyphi B [S. Paratyphi B (dT+)] was investigated. In Germany, the serovar is frequently isolated from broilers. Therefore
D Gopi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 107, 196-202 (2013-02-20)
Hydroxyapatite [HAP, Ca10(PO4)6(OH)2] is the main inorganic component of bone material and is widely used in various biomedical applications due to its excellent bioactivity and biocompatibility. In this paper we have reported the synthesis of hydroxyapatite nanorods by green template
Andrea Bencini et al.
Chemical communications (Cambridge, England), 48(84), 10428-10430 (2012-09-18)
A chiral ditopic polyammonium receptor featuring two [9]aneN(3) moieties separated by a (S)-BINOL linker is able to selectively bind and sense in water (S,S)-tartaric acid over its (R,R)/meso forms.
Preparation of organotrifluoroborate salts: precipitation-driven equilibrium under non-etching conditions.
Alastair J J Lennox et al.
Angewandte Chemie (International ed. in English), 51(37), 9385-9388 (2012-08-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service