Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900609

Sigma-Aldrich

Phthalhydrazide

Vetec, reagent grade, 98%

Synonym(s):

2,3-Dihydro-1,4-phthalazinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N2O2
CAS Number:
Molecular Weight:
162.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

98%

mp

>300 °C (lit.)

SMILES string

O=C1NNC(=O)c2ccccc12

InChI

1S/C8H6N2O2/c11-7-5-3-1-2-4-6(5)8(12)10-9-7/h1-4H,(H,9,11)(H,10,12)

InChI key

KGLPWQKSKUVKMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Schiller et al.
Free radical research, 30(1), 45-57 (1999-04-08)
The reactivity of 5-amino-2,3-dihydro-phthalazine-1,4-dione (luminol) and phthalic hydrazide with hydroxyl radicals was studied. HO*-radicals were generated by the Fenton reaction as well as by water radiolysis. Both luminol and phthalic hydrazide react with hydroxyl radicals under intense chemiluminescence (CL) emission.
I H Hall et al.
The Journal of pharmacy and pharmacology, 41(6), 394-397 (1989-06-01)
The disposition of [14C]2,3-dihydrophthalazine-1,4-dione, a potent hypolipidaemic agent, has been determined after both intravenous and oral administration. Both the routes of administration afforded multi-exponential disposition with an estimated t1/2 of approximately 75 h. After oral administration, the drug was observed
L Butner et al.
International journal of tissue reactions, 18(2-3), 47-55 (1996-01-01)
2,3-Dihydrophthalazine-1,4-diones were observed to be potent anti-inflammatory agents as well as capable of protecting against endotoxin shock in mice at 8 mg/kg i.p. These agents blocked both locally- and centrally-induced pain at 8 mg/kg i.p. In part they appear to
I H Hall et al.
Anti-cancer drugs, 3(1), 55-62 (1992-02-01)
2,3-Dihydrophthalazine-1,4-dione derivatives demonstrated potent cytotoxicity against the growth of murine leukemia cells and human single cell suspension, i.e. Tmolt3 leukemia and HeLa-S3, as well as colon adenocarcinoma and KB nasopharynx. However, only select compounds demonstrated activity against bronchogenic lung, osteosarcoma
I H Hall et al.
Biomedica biochimica acta, 47(4-5), 423-433 (1988-01-01)
2,3-Dihydrophthalazine-1,4-dione effectively lowers serum levels of cholesterol and triglycerides in Sprague Dawley rats after two weeks, after which the cholesterol levels continued to decline. The maximum serum lipid lowering effect on cholesterol or triglyceride levels was during the seventh and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service