Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900563

Sigma-Aldrich

Ethyl stearate

Vetec, reagent grade, 97%

Synonym(s):

Ethyl octadecanoate, Stearic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOC2H5
CAS Number:
Molecular Weight:
312.53
Beilstein:
1788183
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
grade:
reagent grade

grade

reagent grade

product line

Vetec

Assay

97%

bp

213-215 °C/15 mmHg (lit.)

mp

34-38 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCC

InChI

1S/C20H40O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h3-19H2,1-2H3

InChI key

MVLVMROFTAUDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan-Bing Zhang et al.
Journal of Asian natural products research, 8(1-2), 119-123 (2006-06-07)
A new phenolic glycoside (1), mahkoside A, together with six known compounds including mangiferin (2), kaempferol-3-O-beta-D-glucoside (3), dodecanoic acid (4), palmitic acid (5) ethyl stearate (6) and sucrose (7), were isolated from the pit of Mahkota dewa. Their structures were
S Aliaskarisohi et al.
The journal of physical chemistry. B, 115(40), 11631-11637 (2011-08-31)
In a previous work, Muruganathan and Fischer observed laser-induced local collapse of a methyl stearate monolayer. These experiments opened the possibility of studying the collapse mechanism in a highly controlled manner, since the laser intensity can be easily varied and
The dual role of macrophages in the sporozoite-induced malaria infection. A hypothesis.
J P Verhave et al.
International journal of nuclear medicine and biology, 7(2), 149-156 (1980-01-01)
John-Paul Morfin et al.
Alcoholism, clinical and experimental research, 31(2), 265-268 (2007-01-26)
Fatty acid ethyl esters (FAEEs) are generated by the nonoxidative metabolism of alcohol and correlate positively with blood alcohol levels (BAL). As FAEEs are produced predominantly in the liver and bind to albumin in plasma, blood FAEE concentrations may be
M Laposata et al.
Alcoholism, clinical and experimental research, 24(10), 1488-1491 (2000-10-25)
Fatty acid ethyl esters (FAEE) are nonoxidative ethanol metabolites that have been shown to be long term markers of ethanol intake and have been implicated as mediators of ethanol-induced cell injury. Previous studies have indicated that the fatty acid composition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service