Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900539

Sigma-Aldrich

Cinnamaldehyde

Vetec, reagent grade, 93%

Synonym(s):

3-Phenylprop-2-enal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCHO
CAS Number:
Molecular Weight:
132.16
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
grade:
reagent grade

grade

reagent grade

product line

Vetec

Assay

93%

refractive index

n20/D 1.621 (lit.)

bp

248 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

SMILES string

O=C\C=C\c1ccccc1

InChI

1S/C9H8O/c10-8-4-7-9-5-2-1-3-6-9/h1-8H/b7-4+

InChI key

KJPRLNWUNMBNBZ-QPJJXVBHSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

221.0 °F - Pensky-Martens closed cup

Flash Point(C)

105 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnus Bruze et al.
Journal of the American Academy of Dermatology, 48(2), 194-200 (2003-02-13)
Axillary dermatitis is common and overrepresented in individuals with contact allergy to fragrances. Many individuals suspect their deodorants to be the incriminating products. Our aim was to investigate the significance of cinnamic aldehyde in deodorants for the development of axillary
C K Smith et al.
Toxicology and applied pharmacology, 168(3), 189-199 (2000-10-24)
trans-Cinnamaldehyde and trans-cinnamic alcohol have been commonly reported to cause allergic contact dermatitis (ACD) in humans. Cinnamaldehyde is a more potent skin sensitizer than cinnamic alcohol. It has been hypothesized that cinnamic alcohol is a "prohapten" that requires metabolic activation
Michael Bandell et al.
Neuron, 41(6), 849-857 (2004-03-30)
Six members of the mammalian transient receptor potential (TRP) ion channels respond to varied temperature thresholds. The natural compounds capsaicin and menthol activate noxious heat-sensitive TRPV1 and cold-sensitive TRPM8, respectively. The burning and cooling perception of capsaicin and menthol demonstrate
Wen-Xian Du et al.
Journal of agricultural and food chemistry, 60(32), 7799-7804 (2012-07-27)
The addition of plant essential oils to edible films and coatings has been shown to protect against bacterial pathogens and spoilage while also enhancing sensory properties of foods. This study evaluated the effect of adding 0.5 and 0.75% carvacrol (active
Yaoqi Tian et al.
Carbohydrate polymers, 91(2), 586-589 (2012-11-06)
In this study, the high-amylose corn starch-cinnamaldehyde inclusion complex was prepared by an ultrasound treatment and its releasing characteristic was investigated. The results showed that the ultrasound treatment (35°C, 10min and 250W) generated a higher encapsulation rate of 40.2% than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service