Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900490

Sigma-Aldrich

CHAPSO

Vetec, reagent grade, ≥98%

Synonym(s):

3-([3-Cholamidopropyl]dimethylammonio)-2-hydroxy-1-propanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H58N2O8S
CAS Number:
Molecular Weight:
630.88
Beilstein:
5842642
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

grade

reagent grade

description

zwitterionic

product line

Vetec

Assay

≥98%

mol wt

micellar avg mol wt 7000

aggregation number

11

CMC

8 mM (20-25°C)

mp

184-186 °C (lit.)

transition temp

cloud point 90 °C

SMILES string

[H][C@@]12C[C@H](O)CC[C@]1(C)[C@@]3([H])C[C@H](O)[C@]4(C)[C@]([H])(CC[C@@]4([H])[C@]3([H])[C@H](O)C2)[C@H](C)CCC(=O)NCCC[N+](C)(C)CC(O)CS([O-])(=O)=O

InChI

1S/C32H58N2O8S/c1-20(7-10-29(39)33-13-6-14-34(4,5)18-23(36)19-43(40,41)42)24-8-9-25-30-26(17-28(38)32(24,25)3)31(2)12-11-22(35)15-21(31)16-27(30)37/h20-28,30,35-38H,6-19H2,1-5H3,(H-,33,39,40,41,42)/t20-,21+,22-,23?,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

GUQQBLRVXOUDTN-XOHPMCGNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A nondenaturing zwitterionic detergent with characteristics similar to CHAPS, although it is more soluble due to a more polar head group. Useful for the solubilization of integral membrane proteins.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Czerski et al.
Analytical biochemistry, 284(2), 327-333 (2000-08-31)
Bicelles are bilayered discoidal lipid-detergent assemblies which are useful as model membranes. To date, there has been no direct demonstration of functional viability for an integral membrane protein reconstituted into bicelles. In this contribution, the catalytic activity of diacylglycerol kinase
Rachna Ujwal et al.
Journal of visualized experiments : JoVE, 9(59), e3383-e3383 (2012-01-20)
Membrane proteins (MPs) play a critical role in many physiological processes such as pumping specific molecules across the otherwise impermeable membrane bilayer that surrounds all cells and organelles. Alterations in the function of MPs result in many human diseases and
Salem Faham et al.
Protein science : a publication of the Protein Society, 14(3), 836-840 (2005-02-04)
We showed previously that high-quality crystals of bacteriorhodopsin (bR) from Halobacterium salinarum can be obtained from bicelle-forming DMPC/CHAPSO mixtures at 37 degrees C. As many membrane proteins are not sufficiently stable for crystallization at this high temperature, we tested whether
Jenny Frånberg et al.
Biochemistry, 46(25), 7647-7654 (2007-06-01)
Gamma-secretase is important for the development of Alzheimer's disease, since it is a crucial enzyme for the generation of the pathogenic amyloid beta-peptide (Abeta). Most data on gamma-secretase is derived from studies in cell lines overexpressing gamma-secretase components or amyloid
Mako Takami et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(41), 13042-13052 (2009-10-16)
Amyloid beta protein (Abeta), a pathogenic molecule associated with Alzheimer's disease, is produced by gamma-secretase, which cleaves the beta-carboxyl terminal fragment (betaCTF) of beta-amyloid precursor protein in the middle of its transmembrane domain. How the cleavage proceeds within the membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service