Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900454

Sigma-Aldrich

5,5′-Dithiobis(2-nitrobenzoic acid)

Vetec, reagent grade, 98%

Synonym(s):

3-Carboxy-4-nitrophenyl disulfide, 6,6′-Dinitro-3,3′-dithiodibenzoic acid, Bis(3-carboxy-4-nitrophenyl) disulfide, DTNB, Ellman’s Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-SC6H3(NO2)CO2H]2
CAS Number:
Molecular Weight:
396.35
Beilstein:
1896821
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

98%

mp

240-245 °C (dec.) (lit.)

SMILES string

OC(=O)c1cc(SSc2ccc(c(c2)C(O)=O)[N+]([O-])=O)ccc1[N+]([O-])=O

InChI

1S/C14H8N2O8S2/c17-13(18)9-5-7(1-3-11(9)15(21)22)25-26-8-2-4-12(16(23)24)10(6-8)14(19)20/h1-6H,(H,17,18)(H,19,20)

InChI key

KIUMMUBSPKGMOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Bertling et al.
Arteriosclerosis, thrombosis, and vascular biology, 32(8), 1979-1990 (2012-04-28)
Staphylococcus aureus can induce platelet aggregation. The rapidity and degree of this correlates with the severity of disseminated intravascular coagulation, and depends on platelet peptidoglycans. Surface-located thiol isomerases play an important role in platelet activation. The staphylococcal extracellular adherence protein
Zrinka Kovarik et al.
Chemical research in toxicology, 28(5), 1036-1044 (2015-04-04)
Exposure to the nerve agent soman is difficult to treat due to the rapid dealkylation of the soman-acetylcholinesterase (AChE) conjugate known as aging. Oxime antidotes commonly used to reactivate organophosphate inhibited AChE are ineffective against soman, while the efficacy of
Anusha M Gopalakrishnan et al.
Antimicrobial agents and chemotherapy, 59(1), 317-325 (2014-10-29)
Artemisinin-based combination therapy (ACT) is the recommended first-line treatment for Plasmodium falciparum malaria. It has been suggested that the cytotoxic effect of artemisinin is mediated by free radicals followed by the alkylation of P. falciparum proteins. The endoperoxide bridge, the
Chengwei Niu et al.
Journal of ethnopharmacology, 164, 301-308 (2015-02-24)
Diosbulbin B (DB) is the main hepatotoxic compound distributed in Dioscorea bulbifera L., which is widely used for the treatment of cancer and thyroid disorders in Asia. Scutellarin (SC) is the main compound in medicinal herb Scutellaria barbata D. Don
Jun Yan et al.
European journal of medicinal chemistry, 95, 220-229 (2015-03-31)
To identify novel multi-target-directed drug candidates for the treatment of cancer, a series of benzoselenazole-stilbene hybrids were synthesised by combining the pharmacophores of resveratrol and ebselen. The biological assay indicated that all of the hybrids exhibited antiproliferative activities against four

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service