Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900439

Sigma-Aldrich

Uracil

Vetec, reagent grade, 98%

Synonym(s):

2,4(1H,3H)-Pyrimidinedione, 2,4-Dihydroxypyrimidine, 2,4-Pyrimidinediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O2
CAS Number:
Molecular Weight:
112.09
Beilstein:
507828
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

mp

>300 °C (lit.)

SMILES string

O=C1NC=CC(=O)N1

InChI

1S/C4H4N2O2/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)

InChI key

ISAKRJDGNUQOIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra M Carvalho et al.
PloS one, 8(3), e58492-e58492 (2013-03-19)
Links between carbohydrate metabolism and virulence in Streptococcus pneumoniae have been recurrently established. To investigate these links further we developed a chemically defined medium (CDM) and standardized growth conditions that allowed for high growth yields of the related pneumococcal strains
Jaunius Urbonavicius et al.
Methods in enzymology, 425, 103-119 (2007-08-04)
Formation of 5-methyluridine (ribothymidine) at position 54 of the T-psi loop of tRNA is catalyzed by site-specific tRNA methyltransferases (tRNA[uracil-54,C5]-MTases). In eukaryotes and many bacteria, the methyl donor for this reaction is generally S-adenosyl-L-methionine (S-AdoMet). However, in other bacteria, like
Jordan J Feld et al.
The New England journal of medicine, 370(17), 1594-1603 (2014-04-12)
The interferon-free combination of the protease inhibitor ABT-450 with ritonavir (ABT-450/r) and the NS5A inhibitor ombitasvir (also known as ABT-267) plus the nonnucleoside polymerase inhibitor dasabuvir (also known as ABT-333) and ribavirin has shown efficacy against the hepatitis C virus
Pietro Andreone et al.
Gastroenterology, 147(2), 359-365 (2014-05-14)
The interferon-free regimen of ABT-450 (a protease inhibitor), ritonavir, ombitasvir (an NS5A inhibitor), dasabuvir (a non-nucleoside polymerase inhibitor), and ribavirin has shown efficacy in patients with hepatitis C virus (HCV) genotype 1b infection-the most prevalent subgenotype worldwide. We evaluated whether
Daisuke Katagiri et al.
Journal of the American Society of Nephrology : JASN, 24(12), 2034-2043 (2013-10-05)
Accumulating evidence of the beyond-glucose lowering effects of a gut-released hormone, glucagon-like peptide-1 (GLP-1), has been reported in the context of remote organ connections of the cardiovascular system. Specifically, GLP-1 appears to prevent apoptosis, and inhibition of dipeptidyl peptidase-4 (DPP-4)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service