Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900412

Sigma-Aldrich

Tricine

Vetec, reagent grade, ≥99%

Synonym(s):

N-[Tris(hydroxymethyl)methyl]glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)3CNHCH2CO2H
CAS Number:
Molecular Weight:
179.17
Beilstein:
1937804
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥99%

form

crystalline powder

useful pH range

7.4-8.8

pKa (25 °C)

8.1

SMILES string

OCC(CO)(CO)NCC(O)=O

InChI

1S/C6H13NO5/c8-2-6(3-9,4-10)7-1-5(11)12/h7-10H,1-4H2,(H,11,12)

InChI key

SEQKRHFRPICQDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tricine is the most used electrophoresis buffer, which has a lower negative charge than glycine. This in turn helps it to migrate faster. Tricine is also used for the resuspension of cell pellets. Moreover, its high ionic strength leads to more movement of ions and less protein movement. This aids to separate low molecular weight proteins in lower percent acrylamide gels. Tricine is also used as a trailing ion that helps in the resolution of small proteins at lower acrylamide concentrations compared to glycine-sodium dodecyl-sulfate polyacrylamide gel electrophoresis (SDS-PAGE) systems.

Application

Buffer component for separation of low molecular weight peptides.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Encyclopedia of Dairy Sciences (2011)
Arpita Gantayet et al.
Biofouling, 29(1), 77-85 (2012-12-06)
The freshwater zebra mussel (Dreissena polymorpha) is a notorious biofouling organism. It adheres to a variety of substrata underwater by means of a proteinaceous structure called the byssus, which consists of a number of threads with adhesive plaques at the
Hermann Schägger
Nature protocols, 1(1), 16-22 (2007-04-05)
Tricine-SDS-PAGE is commonly used to separate proteins in the mass range 1-100 kDa. It is the preferred electrophoretic system for the resolution of proteins smaller than 30 kDa. The concentrations of acrylamide used in the gels are lower than in
Thierry Rabilloud
Journal of proteomics, 73(8), 1562-1572 (2010-04-17)
Electrophoretic separations of proteins are widely used in proteomic analyses, and rely heavily on SDS electrophoresis. This mode of separation is almost exclusively used when a single dimension separation is performed, and generally represents the second dimension of two-dimensional separations.
Christian Nilsson et al.
Electrophoresis, 31(3), 459-464 (2010-02-02)
Totally porous lipid-based liquid crystalline nanoparticles were used as pseudostationary phase for capillary electroseparation with LIF detection of proteins at physiological conditions using unmodified cyclic olefin copolymer capillaries (Topas, 6.7 cm effective length). In the absence of nanoparticles, i.e. in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service