Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900406

Sigma-Aldrich

L-Serine

≥99%, Vetec

Synonym(s):

(S)-2-Amino-3-hydroxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
105.09
Beilstein:
1721404
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

product name

L-Serine, Vetec, reagent grade, ≥99%

grade

reagent grade

product line

Vetec

Assay

≥99%

form

powder

color

white to off-white

mp

222 °C (dec.) (lit.)

SMILES string

N[C@@H](CO)C(O)=O

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1

InChI key

MTCFGRXMJLQNBG-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Precursor of glycine by serine hydroxymethyltransferase.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Hashimoto et al.
Progress in neuro-psychopharmacology & biological psychiatry, 29(5), 767-769 (2005-06-09)
Several lines of evidence suggest that D-serine, an endogenous agonist of the glycine site on the NMDA receptors, might play a role in the pathophysiology of schizophrenia. The purpose of this study was to determine whether levels of D- and
Miyoshi Haruta et al.
Science (New York, N.Y.), 343(6169), 408-411 (2014-01-25)
Plant cells are immobile; thus, plant growth and development depend on cell expansion rather than cell migration. The molecular mechanism by which the plasma membrane initiates changes in the cell expansion rate remains elusive. We found that a secreted peptide
Vanessa A Morais et al.
Science (New York, N.Y.), 344(6180), 203-207 (2014-03-22)
Under resting conditions, Pink1 knockout cells and cells derived from patients with PINK1 mutations display a loss of mitochondrial complex I reductive activity, causing a decrease in the mitochondrial membrane potential. Analyzing the phosphoproteome of complex I in liver and
Cedric Cagliero et al.
Nucleic acids research, 41(12), 6058-6071 (2013-05-02)
To fit within the confines of the cell, bacterial chromosomes are highly condensed into a structure called the nucleoid. Despite the high degree of compaction in the nucleoid, the genome remains accessible to essential biological processes, such as replication and
Hisaaki Shinohara et al.
Science (New York, N.Y.), 344(6185), 760-764 (2014-05-17)
A switchlike response in nuclear factor-κB (NF-κB) activity implies the existence of a threshold in the NF-κB signaling module. We show that the CARD-containing MAGUK protein 1 (CARMA1, also called CARD11)-TAK1 (MAP3K7)-inhibitor of NF-κB (IκB) kinase-β (IKKβ) module is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service