Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900405

Sigma-Aldrich

Tetraethylammonium chloride

Vetec, reagent grade, 98%

Synonym(s):

TEA chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(Cl)
CAS Number:
Molecular Weight:
165.70
Beilstein:
3563247
EC Number:
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

SMILES string

[Cl-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.ClH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

YMBCJWGVCUEGHA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Shibayama et al.
Pharmaceutical research, 32(9), 2937-2949 (2015-03-21)
Unstirred water layers (UWLs) present an unavoidable complication to the measurement of transport kinetics in cultured cells, and the high rates of transport achieved by overexpressing heterologous transporters exacerbate the UWL effect. This study examined the correlation between measured Jmax
Nanomechanics of cation-π interactions in aqueous solution.
Qingye Lu et al.
Angewandte Chemie (International ed. in English), 52(14), 3944-3948 (2013-03-02)
Fernanda S Gravina et al.
PloS one, 9(10), e111307-e111307 (2014-10-23)
The pacemaker mechanisms activating phasic contractions of vaginal and cervical smooth muscle remain poorly understood. Here, we investigate properties of pacemaking in vaginal and cervical tissues by determining whether: 1) functional pacemaking is dependent on the phase of the estrus
Ling Xiao et al.
Circulation research, 112(10), 1310-1322 (2013-03-28)
A chromosomal haplotype producing cardiac overexpression of dipeptidyl peptidase-like protein-6 (DPP6) causes familial idiopathic ventricular fibrillation. The molecular basis of transient outward current (I(to)) in Purkinje fibers (PFs) is poorly understood. We hypothesized that DPP6 contributes to PF I(to) and
Samuel J Goodchild et al.
The Journal of general physiology, 140(5), 495-511 (2012-10-17)
The open state of voltage-gated potassium (Kv) channels is associated with an increased stability relative to the pre-open closed states and is reflected by a slowing of OFF gating currents after channel opening. The basis for this stabilization is usually

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service