Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900375

Sigma-Aldrich

Adenine hemisulfate salt

Vetec, reagent grade, ≥99%

Synonym(s):

6-Aminopurine hemisulfate salt, Adenine sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H5N5 · 1/2H2SO4
CAS Number:
Molecular Weight:
184.17
Beilstein:
3820263
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

biological source

synthetic

grade

reagent grade

product line

Vetec

Assay

≥99%

form

powder

solubility

0.5 M HCl: 10 mg/mL, clear, colorless

SMILES string

OS(O)(=O)=O.Nc1ncnc2[nH]cnc12.Nc3ncnc4[nH]cnc34

InChI

1S/2C5H5N5.H2O4S/c2*6-4-3-5(9-1-7-3)10-2-8-4;1-5(2,3)4/h2*1-2H,(H3,6,7,8,9,10);(H2,1,2,3,4)

InChI key

LQXHSCOPYJCOMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weixiao Yin et al.
Bio-protocol, 8(9), e2839-e2839 (2018-05-05)
Here, we describe a method that can be used to evaluate the function of predicted signal peptides. This method utilizes the yeast Saccharomyces cerevisiae YTK12 strain and pSUC2 vector in which the pSUC2 vector with fused predicted signal peptide is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service