Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900294

Sigma-Aldrich

HEPES hemisodium salt

Vetec, reagent grade, 99%

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid hemisodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) hemisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17.5N2O4S· 0.5Na
CAS Number:
Molecular Weight:
249.30
UNSPSC Code:
12161700

grade

reagent grade

product line

Vetec

Assay

99%

form

powder

useful pH range

6.8-8.2

pKa 

7.5

solubility

water: 0.5 g/mL, clear to very slightly hazy, colorless

SMILES string

[Na+].OCCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O4S.Na/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14;/h11H,1-8H2,(H,12,13,14);/q;+1/p-1

InChI key

RDZTWEVXRGYCFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service