Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900226

Sigma-Aldrich

Potassium tetraborate tetrahydrate

Vetec, reagent grade, 98%

Synonym(s):

Potassium biborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2B4O7 · 4H2O
CAS Number:
Molecular Weight:
305.50
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

form

powder

SMILES string

O.O.O.O.[K+].[K+].[O-]B([O-])OB1OB(OB=O)O1

InChI

1S/B4O7.2K.4H2O/c5-1-8-3-10-4(11-3)9-2(6)7;;;;;;/h;;;4*1H2/q-2;2*+1;;;;

InChI key

FZICZLCGJZRAHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service