Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900210

Sigma-Aldrich

Boric anhydride

Vetec, reagent grade, 98%

Synonym(s):

Boron trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B2O3
CAS Number:
Molecular Weight:
69.62
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

reagent grade

vapor density

>1 (vs air)

product line

Vetec

Assay

98%

form

fine crystals (may contain lumps)

mp

450 °C (lit.)

solubility

water: ~36 g/L at 25 °C
ethanol: soluble at 20 °C

density

2.46 g/mL at 25 °C (lit.)

SMILES string

O=BOB=O

InChI

1S/B2O3/c3-1-5-2-4

InChI key

JKWMSGQKBLHBQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pratik Koirala et al.
The journal of physical chemistry. A, 117(6), 1310-1318 (2012-11-03)
Using density functional theory based calculations, we have systematically studied the equilibrium geometries, relative stabilities, and electronic and magnetic properties of Fe and Mn atoms interacting with a varying number of BO(2) moieties. These clusters are found to exhibit hyperhalogen
Roger F Brown et al.
Journal of biomedical materials research. Part A, 88(2), 392-400 (2008-02-29)
Glasses containing varying amounts of B(2)O(3) were prepared by partially or fully replacing the SiO(2) in silicate 45S5 bioactive glass with B(2)O(3). The effects of the B(2)O(3) content of the glass on its conversion to hydroxyapatite (HA) and on the
Yinling Wang et al.
Analytical biochemistry, 385(1), 184-186 (2008-11-27)
Glassy carbon electrode modified with boron oxide nanoparticles supported on multiwall carbon nanotubes was obtained via a facile approach. The as-prepared modified electrode exhibits excellent electrocatalytic activity toward the redox of glucose in pH 7.0 phosphate buffer solution. The electrochemical
Noelio O Dantas et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 81(1), 140-143 (2011-07-19)
The structural properties of the glass matrix 40SiO(2)·30Na(2)O·1Al(2)O(3)·(29-x)B(2)O(3)·xFe(2)O(3) (mol%), 0.0≤x≤29.0 were studied by X-ray diffraction (XRD), differential thermal analysis (DTA) and Raman and infrared spectroscopy (FT-IR). XRD demonstrated Fe(3)O(4) crystal formation for Fe(2)O(3) concentrations of 29.0 mol%. DTA showed that
T Raghavendra Rao et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 1116-1122 (2011-06-10)
Optical and physical properties of Ni2+ doped 20ZnO+xLi2O+(30-x)Na2O+50B2O3 (5≤x≤25) glasses are carried out at room temperature. Powder XRD pattern of all the glass samples confirms the amorphous nature. Several physical parameters are evaluated for all the glasses with respect to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service