Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900140

Sigma-Aldrich

Magnesium carbonate basic

Vetec, reagent grade

Synonym(s):

Magnesium hydroxide carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCO3
CAS Number:
Molecular Weight:
84.31
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder

concentration

40.0-44.0% (EDTA titration)

SMILES string

[Mg++].O[Mg]O.[O-]C([O-])=O

InChI

1S/CH2O3.2Mg.2H2O/c2-1(3)4;;;;/h(H2,2,3,4);;;2*1H2/q;2*+2;;/p-4

InChI key

UDYRPJABTMRVCX-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service