Skip to Content
MilliporeSigma
All Photos(3)

Documents

V900095

Sigma-Aldrich

Sodium citrate tribasic dihydrate

Vetec, reagent grade, 98%

Synonym(s):

Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10
Beilstein:
6104939
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

form

powder

mp

>300 °C (lit.)

SMILES string

O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3

InChI key

NLJMYIDDQXHKNR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application


  • Deciphering the orthorhombic crystal structure of a novel NEIL1 nanobody with pseudo-merohedral twinning.: This study showcases the application of sodium citrate tribasic dihydrate in crystallography, focusing on its role in stabilizing protein structures for high-resolution analysis, contributing significantly to molecular biology and enzyme mechanism studies (Prakash et al., 2023).

  • The coiled-coil domain of glycosomal membrane-associated Leishmania donovani PEX14: cloning, overexpression, purification and preliminary crystallographic analysis.: Highlights the use of sodium citrate tribasic dihydrate in the purification processes for protein crystallization, enhancing the understanding of protein structures in parasitology and potential therapeutic targets (Pratap et al., 2020).

  • Determination of Mycotoxins in Brown Rice Using QuEChERS Sample Preparation and UHPLC-MS-MS.: Demonstrates the versatility of sodium citrate tribasic dihydrate in sample preparation for mycotoxin analysis, underscoring its importance in food safety and public health sectors (Nhujak et al., 2016).

  • Electroless Cu Plating on Anodized Al Substrate for High Power LED.: Investigates the role of sodium citrate tribasic dihydrate in electroless copper plating, crucial for the fabrication of high-performance electronic and optical devices (Lee et al., 2015).

  • Crystallization and preliminary X-ray analysis of a novel type of lipolytic hydrolase from Bacillus licheniformis.: Explores the application of sodium citrate tribasic dihydrate in the crystallization of enzymes, providing insights into their structure and function, which is essential for biotechnological applications (Kim et al., 2014).

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evgeny Smirnov et al.
ACS nano, 9(6), 6565-6575 (2015-06-04)
Soft or "liquid-liquid" interfaces were functionalized by roughly half a monolayer of mirror-like nanofilms of gold nanoparticles using a precise interfacial microinjection method. The surface coverage of the nanofilm was characterized by ion transfer voltammetry. These gold nanoparticle films represent
Ruben de Vries et al.
Journal of dairy science, 98(12), 8678-8687 (2015-10-12)
The aim of this study was to evaluate the effect of shortening or omitting the dry period of dairy cows on milk casein composition. For this study, we analyzed milk samples of 90 cows with a dry period of 0
Jun Zhao et al.
Nanotechnology, 25(40), 405101-405101 (2014-09-12)
The treatment of liver injuries or hepatocellular carcinoma (HCC) has been hindered by the lack of efficient drug delivery. Even with the help of nanoparticles or other synthetic delivering agents, a large portion of the dose is still sequestered in
Stijn Mertens et al.
Applied and environmental microbiology, 81(23), 8202-8214 (2015-09-27)
Lager beer is the most consumed alcoholic beverage in the world. Its production process is marked by a fermentation conducted at low (8 to 15°C) temperatures and by the use of Saccharomyces pastorianus, an interspecific hybrid between Saccharomyces cerevisiae and
Rasmus I Jølck et al.
Advanced healthcare materials, 3(10), 1680-1687 (2014-04-16)
External beam radiation therapy relies on the ability to deliver high radiation doses to tumor cells with minimal exposure to surrounding healthy tissue. Advanced irradiation techniques, including image-guided radiation therapy (IGRT), rely on the ability to locate tumors to optimize

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service