Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900026

Sigma-Aldrich

Calcium fluoride

Vetec, reagent grade, 95%

Synonym(s):

Fluorite, Fluorspar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaF2
CAS Number:
Molecular Weight:
78.07
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
95%
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

95%

form

powder

bp

2500 °C (lit.)

density

3.18 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.1%

SMILES string

F[Ca]F

InChI

1S/Ca.2FH/h;2*1H/q+2;;/p-2

InChI key

WUKWITHWXAAZEY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M ten Cate
European journal of oral sciences, 105(5 Pt 2), 461-465 (1997-12-12)
Low concentrations of fluoride have a beneficial effect on enamel and dentin de- and remineralization. After fluoride treatments, such as topical applications, rinses or dentifrices, salivary fluoride concentrations decrease exponentially in a biphasic manner to very low concentrations within a
Y Kitasako et al.
Journal of dental research, 91(4), 370-375 (2012-02-18)
The aim of this study was to assess the effect of chewing gum containing phosphoryl oligosaccharides of calcium (POs-Ca) and a low concentration of fluoride (F) on the hardness of enamel subsurface lesions, utilizing a double-blind, randomized, and controlled in
Nil Ozbek et al.
Food chemistry, 138(1), 650-654 (2012-12-26)
The determination of fluorine in milk samples via the molecular absorption of calcium mono-fluoride (CaF) was performed using a HR-CS-ETAAS. For this purpose, calcium was pipetted to graphite furnace together with samples. The amount of Ca and the graphite furnace
Guanying Chen et al.
ACS nano, 6(9), 8280-8287 (2012-08-30)
We describe the development of novel and biocompatible core/shell (α-NaYbF(4):Tm(3+))/CaF(2) nanoparticles that exhibit highly efficient NIR(in)-NIR(out) upconversion (UC) for high contrast and deep bioimaging. When excited at ~980 nm, these nanoparticles emit photoluminescence (PL) peaked at ~800 nm. The quantum
Lei Cheng et al.
Dental materials : official publication of the Academy of Dental Materials, 28(5), 573-583 (2012-02-10)
Previous studies have developed calcium phosphate and fluoride releasing composites. Other studies have incorporated chlorhexidine (CHX) particles into dental composites. However, CHX has not been incorporated in calcium phosphate and fluoride composites. The objectives of this study were to develop

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service